Learn More
Thermo Scientific™ Sabouraud Dextrose Agar, Emmons with Chloramphenicol and Cycloheximide
Description
Isolate and cultivate pathogenic yeast and molds from clinical specimens with this selective, ready-to-use medium.
Isolate and cultivate pathogenic yeast and molds from clinical specimens with Remel™ Sabouraud Dextrose Agar, Emmons with Chloramphenicol and Cycloheximide. Sabouraud Dextrose Agar, Emmons is a medium with an increased pH of 7.0 and reduced concentration of dextrose from 40 to 20 g/l2, compared to the original composition described in 1892 by Sabouraud1-2.
- Chloramphenicol inhibits some gram-positive and gram-negative bacteria
- Cycloheximide inhibits rapid growing saprophytic fungi
Specifications
Specifications
| Description | Sabouraud Dextrose Agar, Emmons w/Chloramphenicol, Cycloheximide |
| Format | 20mm x 113mm Tube |
| Product Type | Agar Plate |
| Quantity | 20/Pk. |