Learn More
Thermo Scientific™ Sabouraud Dextrose Agar, Emmons w/Chloramphenicol
Description
Isolate and cultivate pathogenic yeast and molds from clinical specimens with Sabouraud Dextrose Agar (Emmons) w/Chloramphenicol.
Isolate and cultivate pathogenic yeast and molds from clinical specimens with Thermo Scientific™ Remel™ Sabouraud Dextrose Agar (Emmons) w/Chloramphenicol. Sabouraud Dextrose Agar (Emmons) is a modification of the original medium developed by Sabouraud in 18921. Emmons neutralized the pH, increasing it to 7.0, and reduced the concentration of dextrose from 40 to 20 g/l2. Chloramphenicol is added to the medium to make it inhibitory to bacterial overgrowth while allowing isolation of pathogenic fungi3.
Specifications
Specifications
| Description | Sabouraud Dextrose Agar, Emmons w/Chloramphenicol |
| Format | F - slant |
| Product Type | Agar |
| Quantity | 20/Pk. |