Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Sabouraud Dextrose Agar, pH 5.6

Catalog No. R454462 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R454462 500 g
R454464 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R454462 Supplier Thermo Scientific™ Supplier No. R454462
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Remel Sabouraud Dextrose Agar, pH 5.6 is used for the isolation of fungi.

Remel Sabouraud Dextrose Agar, pH 5.6 is used for the isolation of fungi.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: USP
Description Sabouraud Dextrose Agar, pH 5.6
Form Powder
Product Type Agar
Quantity 500 g
Yield For 7.7L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.