Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Salmonella Shigella Agar (Dehydrated)

Catalog No. R454502 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
10 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R454502 500 g
R454506 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R454502 Supplier Thermo Scientific™ Supplier No. R454502
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform selective and differential isolation of Salmonella and partly Shigella spp. from clinical specimens using Salmonella Shigella Agar.

Perform selective and differential isolation of Salmonella and partly Shigella spp. from clinical specimens, including feces, and food samples1 using Thermo Scientific™ Remel™ Salmonella Shigella Agar (Dehydrated). Leifson modified the Deoxycholate-Citrate Agar used for the detection of enteric pathogens2 and added bile salts mixture to inhibit the growth of gram-positive organisms.

Salmonella and Shigella are major causes of bacterial enteric illness with transmission occurring via the fecal-oral route, person-to-person contact, or by the ingestion of contaminated food or water.

Perform selective and differential isolation of Salmonella and partly Shigella spp. from clinical specimens, including feces, and food samples using Salmonella Shigella Agar.

  • Ease of Differentiation: Based on the fermentation of lactose; gram-negative bacilli produce pink to red colonies whereas lactose non-fermenters produce transparent to colorless colonies.
  • Selective: Due to the inclusion of selective inhibitory substances brilliant green, bile salts, thiosulphate, and citrate, most gram-positive and coliform organisms are inhibited.

This medium is used for qualitative procedures. This medium contains bile salts, brilliant green and citrates which acts as selective agents and inhibit gram-positive bacteria and coliforms. Sodium thiosulfate, a sulfur source, and ferric ammonium citrate work as indicators to enable organisms which produce H2S to form black-centered colonies, including some strains of Salmonella. Lactose acts as a differentiator.

 

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: COMPF, USDA.
Description Salmonella Shigella Agar
Form Powder
Product Type Agar
Quantity 500 g

REF R119756 is not intended for IVD use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.