Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Selenite Broth w/Cystine

Catalog No. R07841
Change view
Click to view available options
Quantity:
20 x 9 mL
100 x 9 mL
100 x 10 mL
Format:
15mm x 103mm Tube
16mm x 125mm Tube
Packaging Type:
100/Pk.
20/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Packaging Type
R07841 100 x 10 mL 16mm x 125mm Tube 100/Pk.
R064506 100 x 9 mL 15mm x 103mm Tube 100/Pk.
R064508 20 x 9 mL 15mm x 103mm Tube 20/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R07841 Supplier Thermo Scientific™ Supplier No. R07841
This item is not returnable. View return policy

Isolate Salmonella spp. using selective enrichment broth Selenite Broth with Cystine.

Thermo Scientific™ Remel Selenite Broth w/Cystine is a selective enrichment medium for isolation of Salmonella.

Leifson formulated Selenite Broth for enrichment of Salmonella from sources such as feces, urine, and food. Selenite was found to inhibit coliforms and fecal streptococci, however after 12-18 hours of incubation7 the breakthrough of inhibited strains would occur. Addition of L-cystine as an enrichment agent improves the recovery of Salmonella. Bacteria that reduce selenite produce alkali and increase the pH. The acid produced by lactose fermentation causes a decrease in pH and results in a neutral or slightly decreased pH in the medium. Selenite Cystine Broth is used in epidemiological studies to detect organisms from asymptomatic or convalescent patients8 and for recovery of Salmonella and Shigella when it occurs in low numbers in certain specimens.

  • Recommended by the Food and Drug Administration for detection of Salmonella in food materials
  • Recommended by the American Public Health Association (APHA) for isolation of Salmonella from water, foods, and dairy products4-6
  • Selective medium – Sodium selenite inhibits coliforms, enterococci, and other gram-positive organisms
  • Ready-to-use – Convenience of prepared medium

This medium contains casein peptone that supplies nitrogenous substances and carbon compounds necessary for bacterial growth. L-cystine is the enrichment agent that improves recovery of the organisms. Sodium phosphate maintains a stable pH and lessens the toxicity of the selenite. Selective agent sodium selenite inhibits coliforms, enterococci, and other gram-positive organisms.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP.
Description Selenite Broth w/Cystine
Format 16mm x 125mm Tube
Product Type Broth
Packaging Type 100/Pk.
Quantity 100 x 10 mL

REF R07841, REF R112570, and REF R112571 not intended for IVD use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.