Learn More
Thermo Scientific™ Settling Plate, Sterile, Tryptic Soy Agar (Irradiated)
Catalog No. R111874
Change view
Click to view available options
Quantity:
10/Pk.
20/Pk.
Format:
150mm Plate - Double + Bag
Monoplate - Double + Bag
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R111874 20/Pk. 150mm Plate - Double + Bag
R111870 10/Pk. Monoplate - Double + Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R111874 Supplier Thermo Scientific™ Supplier No. R111874
Add to Cart
Add to Cart
This item is not returnable. View return policy
Isolate and cultivate wide variety of organisms like bacteria, yeast and fungi using Settling Plate Sterile Tryptic Soy Agar (Irradiated).
Isolate and cultivate wide variety of organisms like bacteria, yeast and fungi using Thermo Scientific™ Remel™ Settling Plate Sterile Tryptic Soy Agar (Irradiated). It is listed in Standard Methods for the Examination of Water and Wastewater and the Compendium of Methods for the Microbiological Examination of Foods1,2. It is also recommended for use in Official Methods of Analysis of AOAC International, the International Dairy Federation (IDF), and the United States Department of Agriculture (USDA)3,4. Tryptic Soy Agar slants may be used to cultivate, store, and ship bacterial cultures.
Tryptic Soy Agar is prepared following the standard formula for Soybean-Casein Digest Agar Medium in the United States of Pharmacopeia (USP)5. Tryptic Soy Agar many be supplemented with blood to facilitate the growth of more fastidious bacteria or antimicrobial agents to permit the selection of various microbial groups from mixed flora.
  • Recommended for use in Official Methods of Analysis of AOAC International, IDF, and USDA3,4
  • Ready-to-use – Convenience of prepared medium
This medium contains casein and soy peptones that provide nitrogen, amino acids, and peptides necessary for bacterial growth. Dextrose is a ready source of energy. Sodium chloride supplies essential electrolytes and maintains osmotic equilibrium.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description Settling Plate Sterile Tryptic Soy Agar
Format 150mm Plate - Double + Bag
Product Type Agar Plate
Quantity 20/Pk.
Not intended for IVD use.