Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Standard Methods Agar (Dehydrated)

Catalog No. R454704 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R454704 2.5 kg
R454702 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R454704 Supplier Thermo Scientific™ Supplier No. R454704
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Enumerate microbial plate counts from milk, dairy products, food, water and other specimens of sanitary importance with this dehydrated medium.

Enumerate microbial plate counts from milk, dairy products, food, water and other specimens of sanitary importance with Thermo Scientific™ Remel™ Standard Methods Agar (Dehydrated).

The Agar conforms to formulation specified by The Association of Official Analytical Chemists (AOAC)2, American Public Health Association (APHA), Standard Methods for the Examination of Water and Wastewater3,4,5, Environmental Protection Agency (EPA), United States Department of Agriculture (USDA) and International Diary Federation (IDF) confirms the formulation of the Tryptone Glucose Yeast (TGY) Agar.

Enumerate microbial (bacterial) plate counts using the dehydrated medium Standard Methods Agar.

The milk in the Tryptone Glucose Extract Milk Agar is eliminated to provide a medium free from precipitate, to achieve a clearer background and to produce large colonies. This is the formulation of the Standard Methods Agar. The nutrients necessary for bacterial growth are provided by casein peptone present in the medium. The source of B-complex is yeast extract, and dextrose is the energy source.

 

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: APHA and AOAC
Description Standard Methods Agar
Form Powder
Product Type Agar
Quantity 2.5 kg

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.