Learn More
Thermo Scientific™ Remel™ TAT Broth Base
Catalog No. R454792 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R454792 500 g
1 options
This item is not returnable. View return policy
Catalog No. R454792 Supplier Thermo Scientific™ Supplier No. R454792
Add to Cart
Add to Cart
This item is not returnable. View return policy
Detect gram-negative microorganisms and perform sterility testing of cosmetics, topical drugs, and highly viscous substances using this media.
Detect gram-negative microorganisms and perform sterility testing of cosmetics, topical drugs, and highly viscous substances such as salves and ointments using Thermo Scientific™ Remel™ TAT Broth Base (Dehydrated). TAT Broth is also referred to as Fluid Casein Digest-Soy Lecithin Polysorbate 20 Medium. In 1976, Brummer reported lecithin and polysorbate 20 effectively neutralized chemical disinfectant residues which inhibited the growth of viable microorganisms1. It is recommended by the United States Pharmacopeia for use in microbiological examination of non-sterile products2.
TAT Broth is prepared according to the formulation recommended by the Food and Drug Administration3. TAT Broth is a non-inhibitory medium, used mainly for isolating contaminating bacteria in cosmetics, salves and creams4.
  • Recommended by the United States Pharmacopeia for use in the microbiological examination of non-sterile products2.
  • Lecithin and polysorbate 20 effectively neutralize chemical disinfectant residues which inhibit the growth of viable microorganisms1.
This medium contains casein peptone that provides a source of nitrogen, amino acids, and peptides necessary for bacterial growth. Lecithin and polysorbate 20 present in the medium serve to neutralize preservatives in topical drugs and cosmetics.
TRUSTED_SUSTAINABILITY
Specifications
Description TAT Broth Base (Tripticase Azolectin Tween)
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
For Laboratory Use Only