Learn More
Thermo Scientific™ Remel™ TB Decolorizer
Description
Thermo Scientific™ Remel TB Decolorizer is for TB Kinyoun and Ziehl-Neelsen stains.
- For TB Kinyoun and Ziehl-Neelsen stains
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Quantity | Each |
| Volume (Metric) | 250 mL/Bottle |