Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Tetrathionate Broth

Catalog No. R07160
Change view
Click to view available options
Quantity:
20 x 10 mL
100 x 10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R07160 100 x 10 mL
R07162 20 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R07160 Supplier Thermo Scientific™ Supplier No. R07160
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selectively isolate Salmonella from fecal specimens using Tetrathionate Broth enrichment medium.

Selectively isolate Salmonella from fecal specimens using Thermo Scientific™ Remel™ Tetrathionate Broth enrichment medium. This broth was originally described by Muller for cultivation of Salmonella spp1. He demonstrated coliforms were inhibited in the medium while certain enteric gram-negative bacilli, such as Salmonella spp. grew very well. Kauffmann reported an increase in the recovery rate of Salmonella in the Muller medium2,3.

Salmonella are a group of closely related rod-shaped, gram-negative bacteria. Salmonella typically live in animal and human intestines and are shed through feces. Humans become infected most frequently through contaminated water or food. The most common symptoms of Salmonella infection are diarrhea, fever, abdominal cramps, and vomiting.

Tetrathionate broth, when supplemented with Iodine for Tetrathionate, is a selective enrichment medium that is recommended for isolation of Salmonella from fecal specimens.

  • Ready-to-use – The convenience of prepared media.
  • Selective – Bile salts and sodium thiosulfate are selective agents which inhibit gram-positive organisms and some enteric gram-negative bacilli.
  • Formulated as per specifications of the American Public Health Association (APHA), the Association of Official Analytical Chemists (AOAC), the United States Department of Agriculture (USDA), and the United States Pharmacopeia (USP).4-7

This medium contains casein and meat peptones that provide nutritious sources of nitrogen, amino acids, and peptides necessary for bacterial growth. Tetrathionate is formed by the addition of a solution containing iodine and iodide. Organisms capable of reducing tetrathionate, such as Salmonella, flourish in the medium while other fecal organisms are inhibited. Bile salts and sodium thiosulfate are selective agents which inhibit gram-positive organisms and some enteric gram-negative bacilli.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AOAC, BAM, COMPF, SMWW, USDA, USP.
Description Tetrathionate Broth
Format E - 10mL
Product Type Broth
Quantity 100 x 10 mL

Requires addition of Iodine Solution, REF R21228, REF R114350, or REF R114351. REF R112634 and REF R112630 not intended for IVD use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.