Learn More
Thermo Scientific™ Thioglycollate Medium w/Dextrose
Description
Cultivate aerobes and anaerobes using general purpose Thioglycollate Medium w/Dextrose.
Cultivate aerobes and anaerobes using Thermo Scientific™ Remel™ Thioglycollate Medium w/Dextrose. This general-purpose medium was first described by Brewer in 19401 and consists of a small amount of agar and sodium thioglycolate, a reducing substance, to initiate the growth of anaerobes.
Thioglycollate Medium w/Dextrose is a liquid medium recommended for use as a general purpose media. Sodium thioglycollate is the reducing agent which removes molecular oxygen from the medium and prevents the accumulation of peroxides which may be letha
Specifications
Specifications
| Description | Thioglycollate Medium; 18mL; 20 x 113mm Tube |
| Format | 20mm x 113mm Tube |
| Product Type | Microbiology Media |
| Quantity | 100 x 18 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.