Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Thioglycollate Medium w/Dextrose

Catalog No. R08858
Change view
Click to view available options
Quantity:
100 x 9 mL
20 x 10 mL
20 x 18 mL
100 x 10 mL
100 x 18 mL
Format:
15mm x 103mm Tube
16mm x 125mm Tube
20mm x 113mm Tube
F - 18mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R08858 20 x 18 mL F - 18mL
R07176 100 x 10 mL 16mm x 125mm Tube
R07178 20 x 10 mL 16mm x 125mm Tube
R08856 100 x 18 mL 20mm x 113mm Tube
R064700 100 x 9 mL 15mm x 103mm Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. R08858 Supplier Thermo Scientific™ Supplier No. R08858
This item is not returnable. View return policy

Cultivate aerobes and anaerobes using general purpose Thioglycollate Medium w/Dextrose.

Cultivate aerobes and anaerobes using Thermo Scientific™ Remel™ Thioglycollate Medium w/Dextrose. This general-purpose medium was first described by Brewer in 19401 and consists of a small amount of agar and sodium thioglycolate, a reducing substance, to initiate the growth of anaerobes.

Thioglycollate Medium w/Dextrose is a liquid medium recommended for use as a general purpose media. Sodium thioglycollate is the reducing agent which removes molecular oxygen from the medium and prevents the accumulation of peroxides which may be letha

TRUSTED_SUSTAINABILITY

Specifications

Description Thioglycollate Medium w/Dextrose
Format F - 18mL
Product Type Microbiology Media
Quantity 20 x 18 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.