Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Thioglycollate Medium w/Dextrose, Hemin, Vitamin K
GSA_VA

Catalog No. R064722
Change view
Click to view available options
Quantity:
20 x 5 mL
20 x 9 mL
100 x 5 mL
100 x 9 mL
20 x 10 mL
100 x 10 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R064722 20 x 9 mL
R07182 20 x 10 mL
R064730 100 x 5 mL
R064720 100 x 9 mL
R07180 100 x 10 mL
R064732 20 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. R064722 Supplier Thermo Scientific™ Supplier No. R064722
This item is not returnable. View return policy

Cultivate aerobes and anaerobes using Thioglycollate Medium w/Dextrose, Hemin, Vitamin K.

Cultivate aerobes and anaerobes using Thermo Scientific™ Remel™ Thioglycollate Medium w/Dextrose, Hemin, Vitamin K. This general-purpose medium was first described by Brewer in 19401, and consists of a small amount of agar and a reducing substance to initiate the growth of anaerobes.

Thioglycollate Medium w/Dextrose is a liquid medium recommended for use as a general purpose media. Sodium thioglycollate is the reducing agent which removes molecular oxygen from the medium and prevents the accumulation of peroxides which may be letha

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Thioglycollate Medium w/Dextrose, Hemin, Vitamin K, 15 x 103mm Tube
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 20 x 9 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.