Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Tryptic Soy Broth

Catalog No. R08943
Change view
Click to view available options
Quantity:
10 x 25 mL
10 x 90 mL
100 x 15 mL
10 x 100 mL
10 x 500 mL
4 x 1000 mL
Format:
20mm x 113mm Tube
BB - 1000mL
BB Septum - 100mL
BB Septum - 500mL
SB - 100mL
SB - 25mL
WMB Lug - 100mL
WMB Screw - 90mL
Packaging Type:
10/Pk.
100/Pk.
4/Pk.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Packaging Type
R08943 100 x 15 mL 20mm x 113mm Tube 100/Pk.
R112728 10 x 90 mL WMB Screw - 90mL 10/Pk.
R112730 10 x 100 mL WMB Lug - 100mL 10/Pk.
R112731 10 x 100 mL SB - 100mL 10/Pk.
R112732 10 x 500 mL BB Septum - 500mL 10/Pk.
R112740 4 x 1000 mL BB - 1000mL 4/Pk.
R112745 10 x 100 mL BB Septum - 100mL 10/Pk.
R112758 10 x 25 mL SB - 25mL 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. R08943 Supplier Thermo Scientific™ Supplier No. R08943
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Thermo Scientific™ Remel Tryptic Soy Broth (TSB) is a general purpose enrichment broth for cultivation and isolation of microorganisms.

Thermo Scientific™ Remel Tryptic Soy Broth (TSB) is a general purpose enrichment broth for cultivation and isolation of microorganisms.

  • Part numbers beginning with R11 are for sterility testing according to USP (United States Pharmacopeia) and for use as general purpose enrichment broth
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP.
Description Tryptic Soy Broth
Format 20mm x 113mm Tube
Product Type Microbiology Media
Packaging Type 100/Pk.
Quantity 100 x 15 mL

REF numbers beginning with R11 are not intended for IVD use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.