Learn More
Thermo Scientific™ Remel™ Tryptic Soy Broth Modified w/Novobiocin
Catalog No. R458634 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2.5 kg
Catalog No. Quantity
R458634 2.5 kg
1 options
Catalog No. R458634 Supplier Thermo Scientific™ Supplier No. R458634
Selective enrichment liquid medium recommended for cultivation of Escherichia coli O157:H7 from food products.
Remel Tryptic Soy Broth Modified w/Novobiocin (TSB, Modified w/Novobiocin) is a selective enrichment liquid medium recommended for cultivation of Escherichia coli O157:H7 from food products.
TRUSTED_SUSTAINABILITY
Specifications
Description Tryptic Soy Broth Modified w/Novobiocin
Form Powder
Product Type Dehydrated Culture Media
Quantity 2.5 kg
For Laboratory Use Only