Learn More
Thermo Scientific™ Remel™ TT Broth Base (Dehydrated)
Catalog No. R454832 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R454832 500 g
1 options
Catalog No. R454832 Supplier Thermo Scientific™ Supplier No. R454832
Add to Cart
Add to Cart
The Remel TT Broth Base (Hajna) Basal medium (Dehydrated) is used for selective enrichment and isolation of Salmonella.
Remel TT Broth Base (Hajna) Basal medium (Dehydrated) is used for selective enrichment and isolation of Salmonella.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: USDA.
Description TT Broth Base (Hajna)
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Requires Iodine Solution, REF R114352. Not intended for IVD use.