Learn More
Thermo Scientific™ Urea Agar
GSA_VA
Catalog No. R065210
Change view
Click to view available options
Quantity:
100/Pk.
20/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R065210 100/Pk.
R065212 20/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R065210 Supplier Thermo Scientific™ Supplier No. R065210
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Detect urease activity and differentiate microorganisms on the basis of urease production using Urea Agar.
Differentiate microorganisms on the basis of urease activity with Thermo Scientific™ Remel™ Urea Agar. Christensen developed this medium to help in differentiation of rapid urea-positive Proteus spp. from other urea-positive enteric gram-negative bacilli1,2. This medium allows for urease activity to be demonstrated in organisms which have more complex nitrogen requirements3. The medium can also be used to determine urease activity of non-enteric gram-negative bacilli, such as Brucella and Bordetella and to differentiate Cryptococcus and other yeasts4,5.
Urea Agar is used to detect rapid urease activity of the Proteae and non-rapid urease activity of some Enterobacteriaceae. Urea Agar is a lightly buffered medium that contains urea and phenol red, a pH indicator. When organisms utilize urea, ammonia is formed which changes the pH of the medium to alkaline.
  • Ready to use – Convenience of prepared media.
  • Easy to interpret – Intense pink-red color development indicates a positive test.
Urea Agar contains urea as a substrate to detect urease activity. Gelatin peptone promotes rapid growth of many enteric gram-negative bacilli, permitting a decrease in incubation time. Dextrose helps eliminate false-negative reactions and stimulates urease activity in those organisms which hydrolyze urea slowly. Sodium chloride provides electrolytes and monopotassium phosphate acts as a buffer. Phenol red is a pH indicator that changes color from pale-yellow to pink-red in an alkaline environment. Agar is the solidifying agent.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: BAM.
Description Urea Agar
Format 15mm x 103mm Tube - Slant
Quantity 100/Pk.