Learn More
Thermo Scientific™ Remel™ Violet Red Bile Agar w/MUG
Catalog No. R455292 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R455292 500 g
1 options
This item is not returnable. View return policy
Catalog No. R455292 Supplier Thermo Scientific™ Supplier No. R455292
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Remel Violet Red Bile Agar w/MUG is used for the detection of coliforms and the fluorogenic detection of Escherichia coli.
Remel Violet Red Bile Agar w/MUG is used for the detection of coliforms and the fluorogenic detection of Escherichia coli.
TRUSTED_SUSTAINABILITY
Specifications
Description Violet Red Bile Agar w/MUG
Form Powder
Product Type Agar
Quantity 500 g
For Laboratory Use Only