Learn More
Description
Remel XLD Agar is used for the isolation and differentiation of enteric pathogens.
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: USP |
| Description | XLD Agar |
| Form | Powder |
| Product Type | Agar |
| Quantity | 500 g |
| Yield | For 9.0L medium |
For Laboratory Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.