Learn More
Thermo Scientific™ Remel™ XLT4 Agar Base
Catalog No. R459802 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R459802 500 g
1 options
Catalog No. R459802 Supplier Thermo Scientific™ Supplier No. R459802
Remel XLT4 Agar Base is used with Tergitol 4 for the isolation of non-typhi Salmonella species.
Remel XLT4 Agar Base is used with Tergitol 4 for the isolation of non-typhi Salmonella species.
Also available: Tergitol 4, Part No. R450880.
TRUSTED_SUSTAINABILITY
Specifications
Description XLT4 Agar Base
Form Powder
Product Type Agar
Quantity 500 g
For Laboratory Use Only