Learn More
Thermo Scientific™ Remel™ Yeast Nitrogen Base
Catalog No. R459952 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R459952 500 g
1 options
This item is not returnable. View return policy
Catalog No. R459952 Supplier Thermo Scientific™ Supplier No. R459952
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermo Scientific™ Remel Yeast Nitrogen Base is used for carbohydrate assimilation testing of yeast.
Thermo Scientific™ Remel Yeast Nitrogen Base is used for carbohydrate assimilation testing of yeast.
TRUSTED_SUSTAINABILITY
Specifications
Description Yeast Nitrogen Base
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
For Laboratory Use Only