Learn More
Thermo Scientific™ Remel™ YEDC Agar
Catalog No. R456002 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
R456002 500 g
1 options
This item is not returnable. View return policy
Catalog No. R456002 Supplier Thermo Scientific™ Supplier No. R456002
Add to Cart
Add to Cart
This item is not returnable. View return policy
Remel YEDC Agar (Yeast Extract, Dextrose, Chloramphenicol) is used for enumeration of yeast and molds in dairy products.
Remel YEDC Agar (Yeast Extract, Dextrose, Chloramphenicol) is used for enumeration of yeast and molds in dairy products.
TRUSTED_SUSTAINABILITY
Specifications
Description YEDC Agar (Yeast Extract, Dextrose, Chloramphenicol)
Form Powder
Product Type Agar
Quantity 500 g
For Laboratory Use Only