Learn More
TARGETMOL CHEMICALS INC REMOGLIFLOZIN ETABONATE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T34286
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Remogliflozin etabonate (GSK189075A) is a prodrug of regaliflozin and an inhibitor SGLT2 with Ki values of 1.95 2.14 8.57 and 43.1uM for hSGLT2 rSGLT2 rSGLT1 and hSGLT1 respectively. purity: 99%
Catalog No. 50-258-0270
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.