Learn More
Labconco™ Remotely Controlled Service Fixtures
SDP
Catalog No. 1610532 Shop All Labconco Corporation Products
Change view
Click to view available options
Product Type:
Distilled Water Fixture
Gas Fixture
Oxygen Fixture
Standard Fixture
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
16-105-32 Oxygen Fixture
16-105-30 Gas Fixture
16-105-29 Standard Fixture
16-105-31 Distilled Water Fixture
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 16-105-32 Supplier Labconco™ Supplier No. 4952803
Protector fiber glass fume hoods can accommodate up to eight remotely controlled service fixtures, including two gooseneck faucets.
Specifications
For Use With (Equipment) Protector Hood
Includes 0.25 in. Brass Oxygen Valve, Copper Tubing
Product Type Oxygen Fixture