Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Removable Hose Connection Caps
GREENER_CHOICESDP

Catalog No. 31502299 Shop All Kemtech Products
Change view
Click to view available options
Thread Size:
#14
#18
#50
#7
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size
31-502-299 #50
31-501-981 #14
31-501-982 #18
31-502-300 #7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 31-502-299 Supplier Synthware™ Supplier No. RC50
Add to Cart
Add to Cart

Caps for use with removable hose connectors.

  • For use with removable hose connections
  • Available in a wide variety of sizes

Specifications

Color Red
For Use With (Application) Organic Synthesis
Includes Cap Only
Material Nylon
Product Type Removable Hose Connection
Quantity 1 Ea.
Thread Size #50
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.