Learn More
Essendant Max Force Stain Remover, 12 oz Spray Bottle, 12/Carton
Supplier:  Essendant CDC5703700070CT
Encompass_Preferred
Four-in-one stain fighter tackles the toughest spot-cleaning tasks. Amylase enzymes lift food and drink stains. Solvents dissolve oil. Special polymers and surfactants bind grease and soil to the wash water, ensuring a clean rinse. Protease enzymes target protein-based stains, including grass and blood.
Catalog No. 27-910-14
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.