Learn More
Apexbio Technology LLC Repaglinide 135062-02-1 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A53165.1
Encompass_Preferred
Repaglinide (CAS 135062-02-1) is a small-molecule modulator that acts as a blocker of ATP-sensitive potassium (K sub ATP /sub ) channels specifically binding with high affinity to the sulfonylurea receptor 1 (SUR1) subunit when co-expressed with K sub ir /sub 6 2 (K sub d /sub 0 42 nM) Through inhibition of K sub ATP /sub channel activity repaglinide promotes membrane depolarization and subsequent insulin secretion from pancreatic -cells In vivo studies demonstrate its glucose-lowering (hypoglycaemic) effects supporting its utility in antidiabetic research and in mechanistic investigations of pancreatic K sub ATP /sub channel regulation
Catalog No. 50-000-96957
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.