Learn More
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Sterile
Catalog No. 07201489 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
0.05 mL
0.5 mL
1.25 mL
2.5 mL
5 mL
12.5 mL
25 mL
50 mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
07-201-489 25 mL
07-201-483 0.05 mL
07-201-484 0.5 mL
07-201-485 1.25 mL
07-201-486 2.5 mL
07-201-487 5 mL
07-201-488 12.5 mL
07-201-490 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 07-201-489 Supplier Corning™ Supplier No. 6634
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Sterile provide precise, positive-displacement dispensing of viscous or volatile liquids.
  • Ideal for viscous, dense and volatile liquides
  • Minimum increments printed on the syringe itself
  • Accurately deliver selected dose
  • Made from high quality polypropylene and polyethylene
  • Pyrogen-, ATP-, RNase-, and DNase-free
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Certifications/Compliance ISO8655 ISO11137
Material Polypropylene
Sterility Sterile
For Use With (Application) Dispensing of viscous and volatile liquids
Quantity 25/Pk.
Volume (Metric) 25 mL
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov