Learn More
Research Products International Corp Repetitive Dispenser, Complete with 0.6 ml, 1.5 ml and 6.0 ml Syringes
SDP
Supplier:  Research Products International Corp 8100
Encompass_Preferred
Repetitive dispenser allows you to pipette multiple samples up to 58 times without refilling. Dispenses volumes from 10 ul to 5.0 ml, in rapid succession. The liquid is drawn into a reusable plastic syringe that's easy to insert and remove. It locks in place without bothersome clamps or levels to get in the way. Syringes can be autoclaved (except 60 ml size). This model is lightweight with a slim design and perfect balance for effortless pipetting, either right or left handed. Fatigue is reduced to a minimum even during prolonged use. It's so easy to operate. Just slide the syringe into the holder and select the volume to be dispensed with the volume adjustment dial. Fill syringe with fill lever-then dispense samples. Syringes can be refilled and reused when pipetting the same liquid.
Catalog No. 50-550-138
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.