Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell BW™ Replacement 3-Year Oxygen (O2) Sensor

Catalog No. 19806901
Encompass
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
GasAlert MicroClip X3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-806-901 GasAlert MicroClip X3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-806-901 Supplier Honeywell Supplier No. SRX3P
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Honeywell BW™ Replacement 3-Year Oxygen (O2) Sensor for use with GasAlert MicroClip X3

  • Long Life
  • Improved field reliably
  • Superior environmental performance
  • Enhanced response time in extreme conditions
  • 3 Year Warranty
TRUSTED_SUSTAINABILITY

Specifications

Components Measured Oxygen
For Use With (Equipment) GasAlert MicroClip X3
Includes Sensor
Length (Metric) 5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.