Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Replacement Battery

Catalog No. 14559394 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
eLINE Electronic Pipettes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
14-559-394 eLINE Electronic Pipettes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-559-394 Supplier Sartorius Supplier No. 731009
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Replacement Battery for use with eLINE™ electronic pipets.

  • For use with: eLINE™ Electronic Pipettes
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Color Green
Disposable Reusable
For Use With Pipettes
Material Polypropylene
Product Line eLINE
Sterility Non-sterile
For Use With (Equipment) eLINE Electronic Pipettes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.