Learn More
Description
- Plastic cover snaps in place over battery compartment
Specifications
Specifications
| Product Type | Replacement Battery Cover |
| Color | Yellow |
| For Use With (Equipment) | BMP21-PLUS Label Printer |
| Material | Plastic |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.