Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heathrow Scientific Replacement Daylight Plate Kit for Refractometer

Catalog No. 14127360
Change view
Click to view available options
Model:
BTX-1
BTX-20
BX-1
BX-10
BX-2
BX-20
BX-3
BX-4
BX-50
CTX-1
CTX-2
NDX-2
SGX-1
STX-3
SX-1
SX-2
URX-60
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
14-127-360 CTX-2
14-127-325 BTX-1
14-127-309 BTX-20
14-127-358 BX-1
14-127-344 BX-10
14-127-345 BX-2
14-127-312 BX-20
14-127-347 BX-3
14-127-326 BX-4
14-127-327 BX-50
14-127-359 CTX-1
14-127-346 NDX-2
14-127-310 SGX-1
14-127-311 STX-3
14-127-348 SX-1
14-127-349 SX-2
14-127-328 URX-60
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 14-127-360 Supplier Heathrow Scientific Supplier No. 43000DLCTX2
Add to Cart
Add to Cart

All necessary components included

  • Color: Clear
  • Includes: Ring, hinge, plate, and screw
  • Quantity: 1 Each
  • Warranty: 1 Year

Specifications

Type Daylight Plate
For Use With (Equipment) Refractometer
Model CTX-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.