Learn More
Synthware™ Replacement Double Oblique Bore Stopcock
GREENER_CHOICESDP
Catalog No. 31502017 Shop All Kemtech Products
Change view
Click to view available options
Plug Bore:
2 mm
4 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Bore
31-502-017 4 mm
31-502-018 2 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-502-017 Supplier Synthware™ Supplier No. S211944
Add to Cart
Add to Cart
Double Oblique Bore Stopcock has diagonal, parallel bores to allow connection of single stem to both stems on the opposite side.
Specifications
Type Double Oblique Stopcock
Plug Bore 4 mm
Material Borosilicate Glass
For Use With (Application) Organic Synthesis
For Use With (Equipment) Three-way Tap
Plug Material PTFE