Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Idex Replacement Ferrules for SealTight™ Fittings

Catalog No. 05701376
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10/pk
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-701-376 10/pk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-701-376 Supplier Idex Supplier No. F192X
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Slotted end on ferrule creates second compression zone for a tight seal

Specifications

Material PEEK
For Use With (Equipment) 0.0625 in. O.D. Tubing
Type Ferrule
Color Black
Description 10-32 or M6 coned, for 1/16 in. OD tubing
Quantity 10/pk
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.