Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bacharach™ Replacement Fyrite™ Fluid

Catalog No. 23123026
Change view
Click to view available options
For Use With (Equipment):
Fyrite Gas Analyzer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
23-123026 Fyrite Gas Analyzer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-123026 Supplier Bacharach™ Supplier No. 105060

missing translation for 'validMainframeMsg'

For use in Fyrite Gas Analyzer for measuring and analyzing oxygen.

  • For Oxygen Range: 0 to 21% and 0 to 60%
  • 3 Bottles/Carton

Specifications

Detection Limits 0 to 21% and 0 to 60%
Components Measured Oxygen
For Use With (Equipment) Fyrite Gas Analyzer
Type Replacement Fyrite Fluid
For Use With (Application) Measuring and analyzing oxygen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.