Learn More
Thermo Scientific™ Replacement Spring-Loaded Nut for Exactive GC-Orbitrap GC-MS System
Catalog No. 03251524 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Thermo Scientific Q Exactive, Exactive GC Orbitrap GC-MS System
Catalog No. For Use With (Equipment)
03-251-524 Thermo Scientific Q Exactive, Exactive GC Orbitrap GC-MS System
1 options
Catalog No. 03-251-524 Supplier Thermo Scientific™ Supplier No. 1R1204340020
Add to Cart
Add to Cart
Spring-Loaded Nut
Use the Thermo Scientific™ GC-Orbitrap™ High Temperature Nut and Ferrule Kit to obtain leak-free results at high temperature transferline settings on your gas chromatography (GC) Orbitrap. The high-temperature SCP-5000 Vespel ferrules (included) allow continuous operating transferline temperatures of at least 320°C without ferrule deformation or breakdown (common with standard graphite/Vespel ferrules above 280°C), preventing leaks from forming over an extended period of time. The included spring loaded nut ensures a close fit between nut, ferrule and transferline, further preventing leaks from forming.
High-temperature SCP-5000 Vespel ferrules with 0.4mm ID can be used with most 0.1–0.25mm ID gas chromatography columns. Ferrules must be used in conjunction with High Temperature Adjustable Transferline Nut for GC-Orbitrap (included in kit). Nut and ferrules are for use with the Thermo Scientific™ Q Exactive™ GC Orbitrap™ GC-MS/MS system and the Thermo Scientific™ Exactive™ GC Orbitrap™ GC-MS system.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Thermo Scientific Q Exactive, Exactive GC Orbitrap GC-MS System
Type Replacement Spring-Loaded Nut
Description Replacement GC-Orbitrap Spring-Loaded Nut