Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Replacement Gl-45 Drip Ring
SDPSCI_ED

Catalog No. S89639 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Thread Size (Metric):
GL 45
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
S89639 GL 45
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S89639 Supplier United Scientific™ Supplier No. BM0100S
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

United Scientific™ Replacement Gl-45 Drip Ring for GL45 Media Bottles

  • Replacement pouring/drip rings for GL45 Media Bottles
  • Fits all brands of Glass Media Bottles with GL45 Caps
  • Color: Blue

Specifications

Color Blue
Material Polypropylene
Sterility Non-sterile
Thread Size (Metric) GL 45
Neck Finish M0 mm
Graduated No
For Use With (Equipment) With Media Bottles Needing GL 45
Autoclavable Autoclavable
Shape Round
Disposable Reusable
Product Type Pouring Ring
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.