Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ Replacement Items for Freezer/Mill™ Grinding Vial Sets

Catalog No. 6751C4
Change view
Click to view available options
Material:
Polycarbonate
Stainless Steel (440 C)
For Use With (Equipment):
6761 Vial Set
6771 Vial Set
6801 Large Grinding Vial Set and 6871 Large Cr-Free Grinding Vial Set
6881 Vial Set
Spex SamplePrep Freezer/Mill Cryogenic Grinder
Packaging Quantity:
1 Ea.
2 Pk.
20 Pk.
4 Pk.
Product Type:
Center Cylinder
End Plug
Replacement Impactor
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material For Use With (Equipment) Packaging Quantity Product Type
6751C4 Polycarbonate Spex SamplePrep Freezer/Mill Cryogenic Grinder 4 Pk. -
6801C4 Polycarbonate 6801 Large Grinding Vial Set and 6871 Large Cr-Free Grinding Vial Set 4 Pk. Center Cylinder
6751C20 Polycarbonate Spex SamplePrep Freezer/Mill Cryogenic Grinder 20 Pk. Center Cylinder
6801C20 Polycarbonate 6801 Large Grinding Vial Set and 6871 Large Cr-Free Grinding Vial Set 20 Pk. Center Cylinder
68-81C-20 Polycarbonate 6881 Vial Set 20 Pk. Center Cylinder
6751E Stainless Steel (440 C) Spex SamplePrep Freezer/Mill Cryogenic Grinder 2 Pk. End Plug
67-61E - 6761 Vial Set 2 Pk. End Plug
67-71E - 6771 Vial Set 2 Pk. End Plug
6751P Stainless Steel (440 C) Spex SamplePrep Freezer/Mill Cryogenic Grinder 1 Ea. Replacement Impactor
67-61P - 6761 Vial Set 1 Ea. Replacement Impactor
67-71P - 6771 Vial Set 1 Ea. Replacement Impactor
68-81P - - 1 Ea. Replacement Impactor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 6751C4 Supplier Cole-Parmer™ Supplier No. 6751C4
Add to Cart
Add to Cart

Keep spare items to ensure there is no down time.

  • Suitable for use with the Cole-Parmer™ Freezer/Mill™ grinding vial sets
  • Optimize performance of your Cole-Parmer™ Freezer/Mill™ grinding vial sets with a wide range of grinding ball options suitable for your application.

Specifications

For Use With (Equipment) Spex SamplePrep Freezer/Mill Cryogenic Grinder
Material Polycarbonate
Model Cole-Parmer Freezer/Mill
Packaging Quantity 4 Pk.
Sample Volume (English) 0.01 to 0.13 oz.
Sample Volume (Metric) 0.5 to 4 mL
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.