Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell BW™ Replacement MICROceL Sensor

Catalog No. 19806902
Encompass
Encompass_Preferred
Change view
Click to view available options
Components Measured:
Hydrogen Sulfide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
19-806-902 Hydrogen Sulfide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-806-902 Supplier Honeywell BW™ Supplier No. SRHMC
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Honeywell BW™ Replacement MICROceL Sensor for use with GasAlertMax XT II and GasalertMicroClip XT gas detectors

  • 0 to 200 ppm measuring range
  • 1 ppm resolution
TRUSTED_SUSTAINABILITY

Specifications

Components Measured Hydrogen Sulfide
For Use With (Equipment) GasAlertMicroClip and GasAlertMax XT II
Includes Sensor
Length (Metric) 2.54 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.