Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell BW™ Replacement Oxygen (O2) Sensor

Catalog No. 19806900
Encompass
Encompass_Preferred
Change view
Click to view available options
Components Measured:
Oxygen
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
19-806-900 Oxygen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-806-900 Supplier Honeywell Supplier No. SRX2V
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Honeywell BW™ Replacement Oxygen (O2) Sensor for GasAlert Extreme, GasAlertMicroClip XL, GasAlertQuattro, GasAlertMaxXT II, GasAlertMicro 5 Series

TRUSTED_SUSTAINABILITY

Specifications

Components Measured Oxygen
For Use With (Equipment) GasAlert Extreme, GasAlertMicroClip XL, GasAlertQuattro, GasAlertMaxXT II, GasAlertMicro 5 Series
Includes Sensor
Length (Metric) 5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.