Learn More
Drummond™ Replacement Parts Kits for Drummond Microdispensers
Description
Replacement Parts Kits. For Drummond Digital and Dialamatic Microdispensers.
Order Info
For contents of each kit, see chart.
Specifications
Specifications
| Type | Replacement Parts Kit |
| Volume (Metric) | 10 to 50μL |
| For Use With (Equipment) | Drummond Microdispenser |
| Includes | 1 Plunger, 2 O-rings, 1 collet, 1 coil spring, 3 PTFE tip, 1 spring guide |