Learn More
Drummond™ Replacement Parts Kits for Drummond Microdispensers
Catalog No. 211719
Change view
Click to view available options
Volume (Metric):
10 to 50μL
100 to 1000μL
2 to 10μL
20 to 100μL
5 to 25μL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
21-171-9 20 to 100μL
21-171-6 2 to 10μL
21-171-7 5 to 25μL
21-171-8 10 to 50μL
22-231647 100 to 1000μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 21-171-9 Supplier Drummond™ Supplier No. 3000075K
Add to Cart
Add to Cart
Replacement Parts Kits. For Drummond Digital and Dialamatic Microdispensers.
Order Info
For contents of each kit, see chart.
Specifications
Type Replacement Parts Kit
Volume (Metric) 20 to 100μL
For Use With (Equipment) Drummond Microdispenser
Includes 1 Plunger, 2 O-rings, 1 collet, 1 coil spring, 3 PTFE tip, 1 spring guide