Learn More
Nilfisk™ Replacement Parts for Nilfisk™ Vacuums
Catalog No. 19033526 Shop All Nilfisk of America Products
Change view
Click to view available options
Product Type:
Dust Bag
Polycomposite Microfilter
Vacuum Hose
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-033-526 Vacuum Hose
19-033-530 Polycomposite Microfilter
19-033-529 Dust Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-033-526 Supplier Nilfisk™ Supplier No. 140 8474 000
Add to Cart
Add to Cart
For use with Nilfisk-Advance Vacuums
  • Disposable bags sold in packs of five
  • AES microfilter and HEPA cartridge sold individually
Specifications
For Use With (Application) For Vacuuming in Cleanrooms, Pharmaceutical Labs, and other Clean Environments
Product Type Vacuum Hose
For Use With (Equipment) Nilfisk-advance Vacuum
Material Plastic, Stainless Steel
Length (English) 10 ft.
Length (Metric) 3.04 m