Learn More
Thermo Scientific™ Replacement Plungers for Syringes
Catalog No. 365RK351
Change view
Click to view available options
For Use With (Equipment):
Thermo 36500500
Thermo 365G2111 or 365G2161
Thermo 365H3771
Thermo 365I2561 or 365I2611
Thermo 365J2831 or 365J2881
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
36-5RK-351 Thermo 36500500
36-5RP-451 Thermo 365G2111 or 365G2161
36-5RP-471 Thermo 365H3771
36-5RP-481 Thermo 365I2561 or 365I2611
36-5RP-491 Thermo 365J2831 or 365J2881
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 36-5RK-351 Supplier Thermo Scientific™ Supplier No. 365RK351
Thermo Scientific™ Replacement Plungers for Syringes are available for selected syringes from a variety of manufacturers.
Thermo Scientific™ Replacement Plungers for Syringes are available for selected syringes from a variety of manufacturers.
TRUSTED_SUSTAINABILITY
Specifications
Description For use with 365D1611
Needle Gauge 23 G
Length (English) Needle 1.65354 in.
Product Type Syringe Plunger
Volume (Metric) 10 μL
For Use With (Equipment) Thermo 36500500