Learn More
Marketlab, Inc. Replacement Ribbon for T-4U Time and Date Recorder
Description
- Compatible with model ML3191
Specifications
Specifications
| Color | Black |
| For Use With (Equipment) | T-4U Time and Date Recorder |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.