Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ Valves and Seals for seripettor™

Catalog No. 13880707 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
Color:
Black
Product Type:
Sealing Washer
Autoclavable:
Autoclavable
Includes:
Five sealing washers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type Autoclavable Includes
13-880-707 Black Sealing Washer Autoclavable Five sealing washers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-880-707 Supplier BRANDTECH™ BRAND™ Supplier No. 6788
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Replacement valves and seals for use with seripettor™ bottletop dispensers.

Filling valve set

  • Includes: 1 x filling valve body, 1 x filling valve, 1 x discharge valve, and 2 x sealing rings

Sealing rings

  • Five per pack
  • Also compatible with the Quiksip™ bottletop aspirator
TRUSTED_SUSTAINABILITY

Specifications

Product Type Sealing Washer
Autoclavable Autoclavable
Color Black
Material EPDM
For Use With (Application) Dispensing liquid
For Use With (Equipment) seripettor; QuikSip
Includes Five sealing washers
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.