Learn More
Synthware™ Replacement Straight Bore Stopcock
GREENER_CHOICESDP
Catalog No. 31502007 Shop All Kemtech Products
Change view
Click to view available options
Plug Bore:
2 mm
4 mm
6 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Bore
31-502-007 4 mm
31-502-006 2 mm
31-502-008 6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-502-007 Supplier Synthware™ Supplier No. S181530
Add to Cart
Add to Cart
Supplied with PTFE plug, one PTFE washer and one O-ring.
Specifications
Type Straight Bore Stopcock
Plug Bore 4 mm
Material Borosilicate Glass
For Use With (Application) Organic Synthesis
For Use With (Equipment) Straight Bore Tap
Includes PTFE plug, One PTFE washer, One o-ring
Plug Material PTFE