Learn More
MSA™ Replacement Suspensions for V-Gard™ Caps and Hats
Catalog No. 19039200 Shop All MSA Products
Change view
Click to view available options
Construction:
4-Point webbed suspension, adjustable dual Nylon crown straps, perforated Vinyl sweatband
4-Point webbed suspension, adjustable dual Nylon crown straps, perforated Vinyl sweatband, large easy adjustment knob
Product Type:
Staz-On™ Pin Lock Suspension System
Staz-On™ Suspension System
Size:
Large
Standard
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Construction Product Type Size
19-039-200 4-Point webbed suspension, adjustable dual Nylon crown straps, perforated Vinyl sweatband Staz-On™ Pin Lock Suspension System Standard
18-999-4017 4-Point webbed suspension, adjustable dual Nylon crown straps, perforated Vinyl sweatband, large easy adjustment knob Staz-On™ Suspension System Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-039-200 Supplier MSA™ Supplier No. 10087218
Add to Cart
Add to Cart
Designed for easy clip-in to the V Gard hat or cap shell, users can adjust the liner for a secure and comfortable fit.
Staz-On Suspension
Comfortable four-point webbed suspension features adjustable dual nylon crown straps, a perforated vinyl sweatband with pinlock closure
Fas-Trac Ratchet Suspension
Four-point webbed suspension features adjustable dual nylon crown straps, a perforated vinyl sweatband, and a large easy-adjustment knob closure
1-Touch Suspension
One-handed slide adjustment; easy squeeze opening
Specifications
Construction 4-Point webbed suspension, adjustable dual Nylon crown straps, perforated Vinyl sweatband
For Use With (Equipment) V-Gard Hard Hat and Cap
Includes Headband, nape strap, sweatband and adjustable crown straps
Material Plastic, Nylon
Product Type Staz-On™ Pin Lock Suspension System
Size Standard
Size (Metric) 16.51 to 20.32 cm
Size (English) 6.5 to 8 in.