Learn More
MilliporeSigma™ Supelco™ Replacement Thermogreen™ LB-2 Septa
Catalog No. 111012091 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Pre-conditioned
Catalog No.
11-101-2091 Pre-conditioned
1 options
Catalog No. 11-101-2091 Supplier MilliporeSigma™ Supelco™ Supplier No. 29458U
Add to Cart
Add to Cart
Configured for Gas Sampling Bags with Screw Cap Valves (SCV)
An improved version over the original Thermogreen™ LB-1 Septa
TRUSTED_SUSTAINABILITY
Specifications
Color Green
Description Pre-conditioned
Quantity 10 Pk.
Temperature (Metric) Max 350°C
Type Septa
Thermogreen is a registered trademark of Sigma-Aldrich Co. LLC